missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tetrachloroaurate(III) hydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2329203

 View more versions of this product

Catalog No. AAB2329203



Description

Description

Potassium tetrachloroaurate(III) hydrate is used as primary and secondary intermediates. A Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrachloroaurate(III) hydrate is used as primary and secondary intermediates. A Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.

Solubility
Soluble in water, ethanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Potassium tetrachloroaurate(III) hydrate
27988-75-6
AuCl4K
MFCD00043088
14,7681
Soluble in water,ethanol.
[K+].Cl[Au+3](Cl)(Cl)Cl
377.86
377.88 (Anhydrous)
357°C (decomposition)
1 g
KAuCl4·xH2O
UN3260
potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion
IKHCGJWURGSQRT-UHFFFAOYSA-J
potassium;tetrachlorogold(1-);hydrate
91886546
99%

Chemical Identifiers

27988-75-6
377.86
IKHCGJWURGSQRT-UHFFFAOYSA-J
91886546
[K+].Cl[Au+3](Cl)(Cl)Cl
AuCl4K
MFCD00043088
potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion
potassium;tetrachlorogold(1-);hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 237-190-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only