missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tetrachloroaurate(III), 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC316120050

 View more versions of this product

Catalog No. AC316120050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

377.88
Crystalline Powder
Yellow-orange
Glass bottle
98%
13682-61-6
KAuCl4
≥97.5%
5g
15, 7800
Potassium tetrachloroaurate(III)
AuCl4K
MFCD00043088

Chemical Identifiers

98%
KAuCl4
MFCD00043088
≥97.5%
AuCl4K
377.88
Potassium tetrachloroaurate(III)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.