missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium tetrabromoplatinate(II), 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4554888

 View more versions of this product

Catalog No. AA4554888



Specifications

Specifications

Potassium tetrabromoplatinate (II)
(metals basis)
K2PtBr4
br4pt.2k, dipotassium;tetrabromoplatinum 2-, dipotassium tetrabromoplatinumdiuide
[K+].[K+].Br[Pt-2](Br)(Br)Br
592.897
592.9
99.9%
2.5g
13826-94-3
Br4K2Pt
MFCD00011376
AXZAYXJCENRGIM-UHFFFAOYSA-J
dipotassium;tetrabromoplatinum(2-)
71310129
Crystalline
>300°C

Chemical Identifiers

13826-94-3
592.897
AXZAYXJCENRGIM-UHFFFAOYSA-J
71310129
[K+].[K+].Br[Pt-2](Br)(Br)Br
Br4K2Pt
MFCD00011376
br4pt.2k, dipotassium;tetrabromoplatinum 2-, dipotassium tetrabromoplatinumdiuide
dipotassium;tetrabromoplatinum(2-)
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TP1830000

TSCA : TSCA

SDS Documents