Learn More
Potassium tetrabromoplatinate(II), 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045548ME
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium tetrabromoplatinate (II) | |
13826-94-3 | |
500 mg | |
Br4K2Pt | |
MFCD00011376 | |
AXZAYXJCENRGIM-UHFFFAOYSA-J | |
dipotassium;tetrabromoplatinum(2-) | |
71310129 | |
99.9% |
>300°C | |
Crystalline | |
(metals basis) | |
K2PtBr4 | |
br4pt.2k, dipotassium;tetrabromoplatinum 2-, dipotassium tetrabromoplatinumdiuide | |
[K+].[K+].[Br-].[Br-].[Br-].[Br-].[Pt++] | |
592.90 | |
592.9 |
Chemical Identifiers
13826-94-3 | |
592.90 | |
AXZAYXJCENRGIM-UHFFFAOYSA-J | |
71310129 | |
[K+].[K+].[Br-].[Br-].[Br-].[Br-].[Pt++] |
Br4K2Pt | |
MFCD00011376 | |
br4pt.2k, dipotassium;tetrabromoplatinum 2-, dipotassium tetrabromoplatinumdiuide | |
dipotassium;tetrabromoplatinum(2-) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 237-536-1
RTECSNumber : TP1830000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only