missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tetraborate tetrahydrate, 99.5%, extra pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC259570051

 View more versions of this product

Catalog No. AC259570051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassiumtetraboratetetrahydrate
99.5%
K2B4O7·4H2O
Potassium biborate tetrahydrate
305.5
Powder
Extra Pure
20ppm max.
20ppm max.
Solubility in water: soluble.
White
5kg
12045-78-2
B4K2O7·4H2O
14,768
Solubility in water: soluble.
305.5
99.5%
20ppm max
20ppm max.
Plastic drum
500ppm max.
850.0°C

Chemical Identifiers

12045-78-2
B4K2O7·4H2O
305.5
Potassium biborate tetrahydrate
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging the unborn child.

GHS P Statement
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

SDS Documents