missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium (tert-butoxymethyl)trifluoroborate, 97%, ACROS Organics™

Chemical Identifiers

CAS 910251-10-4
Molecular Formula C5H11BF3KO
Molecular Weight (g/mol) 194.046
InChI Key XBEPCFNGTDJTIY-UHFFFAOYSA-N
Synonym potassium tert-butoxymethyl trifluoroborate, potassium tert-butoxymethyltrifluoroborate, potassium tert-butoxymethyl trifluoroboranuide, potassium tert-butoxy methyl trifluoroboranuide, potassium tert-butoxymethyl trifluoroborate, potassium tert-butoxymethyl trifluoro borate 1-
PubChem CID 45588181
IUPAC Name potassium;trifluoro-[(2-methylpropan-2-yl)oxymethyl]boranuide
SMILES [B-](COC(C)(C)C)(F)(F)F.[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444840010 SDS View Documents Acros Organics
AC444840010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

910251-10-4
194.046
potassium tert-butoxymethyl trifluoroborate, potassium tert-butoxymethyltrifluoroborate, potassium tert-butoxymethyl trifluoroboranuide, potassium tert-butoxy methyl trifluoroboranuide, potassium tert-butoxymethyl trifluoroborate, potassium tert-butoxymethyl trifluoro borate 1-
potassium;trifluoro-[(2-methylpropan-2-yl)oxymethyl]boranuide
C5H11BF3KO
XBEPCFNGTDJTIY-UHFFFAOYSA-N
45588181
[B-](COC(C)(C)C)(F)(F)F.[K+]

Specifications

1g
97%
potassium tert-butoxymethyl trifluoroborate, potassium tert-butoxymethyltrifluoroborate, potassium tert-butoxymethyl trifluoroboranuide, potassium tert-butoxy methyl trifluoroboranuide, potassium tert-butoxymethyl trifluoroborate, potassium tert-butoxymethyl trifluoro borate 1-
[B-](COC(C)(C)C)(F)(F)F.[K+]
194.046
194.05
910251-10-4
C5H11BF3KO
XBEPCFNGTDJTIY-UHFFFAOYSA-N
potassium;trifluoro-[(2-methylpropan-2-yl)oxymethyl]boranuide
45588181
Potassium (tert-butoxymethyl)trifluoroborate