Please login to your online account to display your discounted pricing

Potassium (tert-butoxymethyl)trifluoroborate, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC444840050

 View more versions of this product

Catalog No. AC444840050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 194.05
Quantity 1g
Chemical Name or Material Potassium (tert-butoxymethyl)trifluoroborate
Assay Percent Range 97%
Molecular Formula C5H11BF3KO
CAS 910251-10-4

Specifications

Chemical Name or Material Potassium (tert-butoxymethyl)trifluoroborate
Assay Percent Range 97%
Molecular Formula C5H11BF3KO
Formula Weight 194.05