Please login to your online account to display your discounted pricing

Potassium tert-butoxide, pure, 1.7M (20 wt.%) solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC428791000

 View more versions of this product

Catalog No. AC428791000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 112.21
Physical Form Solution
Quantity 100mL
Color Pale yellow to amber
Packaging AcroSeal™ Glass Bottle
Purity Grade Pure
Density 0.9290g/mL
Concentration 1.60 to 1.75M
Synonym Potassium tert-butylate
CAS 109-99-9
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. May form explosive peroxides.
CAS 865-47-4
Chemical Name or Material Potassium tert-butoxide
Name Note Pure, 1.7M (20 wt.%) solution in THF
MDL Number MFCD00012162
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present an

Specifications

Chemical Name or Material Potassium tert-butoxide
Name Note Pure, 1.7M (20 wt.%) solution in THF
CAS 865-47-4
CAS 109-99-9
Purity Grade Pure
Molecular Formula C4H9KO
Linear Formula (CH3)3COK
Formula Weight 112.21
MDL Number MFCD00012162
Synonym Potassium tert-butylate
Accessories

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present an

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. May form explosive peroxides.

SDS & Safety Documents