missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tert-Butoxide (12% in Tetrahydrofuran, ca. 1mol/L), TCI America™

Manufacturer:  TCI America P1619500ML

 View more versions of this product

Catalog No. P1619500ML



Specifications

Specifications

Potassium tert-Butoxide (12% in Tetrahydrofuran, ca. 1mol/L)
C4H9KO
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium;2-methylpropan-2-olate
23665647
Clear Liquid-Slightly Cloudy Liquid at 20°C
500mL
865-47-4
2924
CC(C)(C)[O-].[K+]
112.213
112.21
Yellow

Chemical Identifiers

865-47-4
112.213
23665647
CC(C)(C)[O-].[K+]
C4H9KO
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium;2-methylpropan-2-olate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3100

TSCA : Yes

Documents