missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tert-Butoxide (12% in Tetrahydrofuran, ca. 1mol/L), TCI America™

Manufacturer:  TCI America P1619100ML

 View more versions of this product

Catalog No. P1619100ML



Specifications

Specifications

Potassium tert-Butoxide (12% in Tetrahydrofuran, ca. 1mol/L)
100mL
C4H9KO
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
CC(C)(C)[O-].[K+]
112.213
112.21
Yellow
865-47-4
2924
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium;2-methylpropan-2-olate
23665647
Clear Liquid-Slightly Cloudy Liquid at 20°C

Chemical Identifiers

865-47-4
112.213
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
potassium;2-methylpropan-2-olate
C4H9KO
LPNYRYFBWFDTMA-UHFFFAOYSA-N
23665647
CC(C)(C)[O-].[K+]
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3100

TSCA : Yes

Documents