Please login to your online account to display your discounted pricing

Potassium superoxide, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  355420250

 View more versions of this product

Catalog No. AC355420250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder or Chunks
Formula Weight 71.1
Quantity 25g
Color Yellow
RTECS Number TT6053000
EINECS Number 234-746-5
TSCA TSCA
Packaging Plastic bottle
Molecular Formula KO2
MDL Number MFCD00082411
CAS 12030-88-5
Synonym Potassium dioxide
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Potassium superoxide
Linear Formula KO
Additional Information O2-content:> 215 cc/g

Specifications

Chemical Name or Material Potassium superoxide
CAS 12030-88-5
Molecular Formula KO2
Linear Formula KO
Formula Weight 71.1
MDL Number MFCD00082411
Synonym Potassium dioxide
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.
Reacts violently with water.

SDS & Safety Documents