missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium sulfate, Puratronic™, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 1084506

 View more versions of this product

Catalog No. AA1084506



Specifications

Specifications

Potassium sulfate
(metals basis)
K2SO4
14,7674
Soluble in water and glycerol. Insoluble in alcohol,acetate and carbon disulfide.
[O-]S(=O)(=O)[O-].[K+].[K+]
174.253
CHEBI:32036
Powder
Puratronic™
Odorless
White
5g
7778-80-5
K2O4S
MFCD00011388
arcanum duplicatum, caswell no. 702, dipotassium sulfate, kalium sulphuricum, potassium sulfate, potassium sulphate, sal polychrestum, sulfuric acid dipotassium salt, sulfuric acid, dipotassium salt, sulfuric acid, potassium salt
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium;sulfate
24507
174.27
99.997%
2.662 g/mL
1689°C
1,069°C

Chemical Identifiers

7778-80-5
174.253
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
dipotassium;sulfate
K2O4S
MFCD00011388
arcanum duplicatum, caswell no. 702, dipotassium sulfate, kalium sulphuricum, potassium sulfate, potassium sulphate, sal polychrestum, sulfuric acid dipotassium salt, sulfuric acid, dipotassium salt, sulfuric acid, potassium salt
CHEBI:32036
[O-]S(=O)(=O)[O-].[K+].[K+]
Safety and Handling

Safety and Handling

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-915-5

RTECSNumber : TT5900000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents