Please login to your online account to display your discounted pricing

Potassium Sulfate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$286.09 - $1,266.36

Chemical Identifiers

Chemical Name or Material Potassium Sulfate
CAS 7778-80-5
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula K2SO4
Formula Weight 174.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003837

 
avantor performance
328205
Poly Bottle 2.5kg Each for $1,266.36

02003838

 
avantor performance
328201
Poly Bottle 500g Each for $286.09
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 174.27
Chemical Name or Material Potassium Sulfate
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 2.66kg
Molecular Formula K2SO4
CAS 7778-80-5
Quantity 2.5kg