missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium sulfate, ACS, 99.0% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 14311A3

 View more versions of this product

Catalog No. AA14311A3



Specifications

Specifications

Potassium sulfate
7778-80-5
K2O4S
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
[O-]S(=O)(=O)[O-].[K+].[K+]
174.253
CHEBI:32036
Crystalline
2kg
For ACS analysis
99.0% min
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium;sulfate
24507
174.27
2.662g/mL

Chemical Identifiers

7778-80-5
174.253
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
dipotassium;sulfate
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
CHEBI:32036
[O-]S(=O)(=O)[O-].[K+].[K+]
Safety and Handling

Safety and Handling

EINECSNumber : 231-915-5

Documents