missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium sulfate, 99+%, for analysis, anhydrous powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 207775000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1689.0°C | |
1067.0°C | |
7778-80-5 | |
6 to 8 (5% soln. at 20°C) | |
500 g | |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
dipotassium;sulfate | |
24507 | |
174.25 | |
Analytical | |
2.66 |
Potassium sulfate | |
p.a. | |
White | |
Very Fine Crystals or Powder | |
99+% | |
K2SO4 | |
15, 7793 | |
Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol | |
[K+].[K+].[O-]S([O-])(=O)=O | |
174.25 | |
CHEBI:32036 | |
≥99% | |
Plastic bottle |
Chemical Identifiers
7778-80-5 | |
174.25 | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
24507 | |
[K+].[K+].[O-]S([O-])(=O)=O |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
CHEBI:32036 |
Safety and Handling
EINECSNumber : 231-915-5
RTECSNumber : TT5900000
TSCA : TSCA
RUO – Research Use Only