Learn More
Potassium sulfate, 99.99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03048514
Description
Potassium sulfate is used in the production of fertilizers (i.e. providing potassium and sulfur), potassium alum, potassium carbonate, and glass. In analytical chemistry, it is used in the determination of nitrogen by Kjeldahl method and protein in foodstuffs. Also, it is used in the electrostatic effects on pKa values of amino acid residues in the staphylococcal nuclease. It is also used in the separation of protein through electrophoresis. It also serves as a laboratory reagent..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1689°C | |
1,069°C | |
Powder | |
(metals basis) | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
dipotassium;sulfate | |
24507 | |
174.27 | |
Odorless |
Potassium sulfate | |
7778-80-5 | |
25 g | |
K2O4S | |
14,7674 | |
Soluble in water and glycerol. Insoluble in alcohol,acetate and carbon disulfide. | |
[K+].[K+].[O-]S([O-])(=O)=O | |
174.25 | |
CHEBI:32036 | |
99.99% | |
2.662 g/mL |
Chemical Identifiers
7778-80-5 | |
174.25 | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
24507 | |
[K+].[K+].[O-]S([O-])(=O)=O |
K2O4S | |
MFCD00011388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
CHEBI:32036 |
Safety and Handling
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 231-915-5
RTECSNumber : TT5900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only