missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$169.43 - $295.09
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.548 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
IUPAC Name | potassium;chloride |
SMILES | [Cl-].[K+] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
K | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7447-40-7 | |
74.548 | |
4873 | |
potassium;chloride |
ClK | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
CHEBI:32588 | |
[Cl-].[K+] |
Specifications
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
7447-40-7,7647-01-0,7732-18-5 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
Assurance | |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
125mL |
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation | |
ClK | |
[Cl-].[K+] | |
74.548 | |
CHEBI:32588 | |
Triple-leached LDPE bottle | |
2% HNO3 | |
Potassium Standard |
Safety and Handling
Causes severe skin burns and eye damage
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only