missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™

$80.74 - $226.46

Chemical Identifiers

CAS 7447-40-7,7647-01-0,7732-18-5
Chemical Name or Material Potassium Standard
Grade Assurance Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
PLK22T SDS Spex Certiprep Llc
PLK22T
Poly Bottle 250mL Each for $226.46
PLK22M Spex Certiprep Llc
SPXPLK22M
Triple-leached LDPE bottle 30mL Each for $80.74
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

7447-40-7,7647-01-0,7732-18-5
Potassium Standard
Assurance Grade

Specifications

7447-40-7,7647-01-0,7732-18-5
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Potassium Standard
Poly Bottle
250mL
Assurance Grade

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient