missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium, solid, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1029906

 View more versions of this product

Catalog No. AA1029906



Specifications

Specifications

Potassium
7440-09-7
0.86g/cm3
K
atom
[K]
Odorless
5462222
Solid
5g
Silver colored solid
99.95%
MFCD00133776
NPYPAHLBTDXSSS-UHFFFAOYSA-N
potassium
39.10
CHEBI:26216
Silver

Chemical Identifiers

7440-09-7
39.10
NPYPAHLBTDXSSS-UHFFFAOYSA-N
5462222
potassium
K
MFCD00133776
atom
CHEBI:26216
[K]
Safety and Handling

Safety and Handling

H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-119-8

Documents