Please login to your online account to display your discounted pricing

Potassium sodium tartrate tetrahydrate, +99%, for analysis, ACROS Organics™


Quantity 5kg
Packaging Plastic Bottle

Manufacturer: thermo fisher scientific  AC202860051

 View more versions of this product

Catalog No. AC202860051

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 282.22
Physical Form Crystals
Quantity 5kg
Color White semi-transparent
Beilstein 03, 525
Merck Index 15, 7790
Packaging Plastic Bottle
Assay 99 to 102%
pH 5.5 to 8.5 (5% soln. at 20°C)
Purity Grade For analysis
Decomposition Information >100°C
Specific Rotation Condition + 22.10 (20.00°C c=10,water)
Lead (Pb) 10ppm max.
Iron (Fe) 10ppm max.
Linear Formula KO2CCH(OH)CH(OH)CO2Na·4H2O
Calcium (Ca) 50ppm max.
Assay Percent Range 99+%
Chemical Name or Material Potassium sodium tartrate tetrahydrate
Molecular Formula C4H4KNaO6·4H2O
MDL Number MFCD00150989
Infrared Spectrum Authentic
Synonym Rochelle salt
Sulfate 50ppm max.
Chlorine (Cl) 50ppm max.
CAS 6381-59-5

Specifications

Chemical Name or Material Potassium sodium tartrate tetrahydrate
CAS 6381-59-5
Purity Grade For analysis
Assay 99 to 102%
Assay Percent Range 99+%
Molecular Formula C4H4KNaO6·4H2O
Linear Formula KO2CCH(OH)CH(OH)CO2Na·4H2O
Formula Weight 282.22
MDL Number MFCD00150989
Synonym Rochelle salt