missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium sodium tartrate tetrahydrate, +99%, for analysis, ACROS Organics™

Manufacturer:  Acros Organics 202865000

 View more versions of this product

Catalog No. AC202865000



Specifications

Specifications

Potassium sodium tartrate tetrahydrate
Authentic
+ 22.10 (20.00°C c=10,water)
500g
6381-59-5
C4H12KNaO10
MFCD00150989
15, 7790
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
2724148
Crystals
Analysis
>100°C
Plastic bottle
White
5.5 to 8.5 (5% soln. at 20°C)
99+%
KO2CCH(OH)CH(OH)CO2Na·4H2O
03, 525
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
282.218
282.22
99 to 102%

Chemical Identifiers

6381-59-5
282.218
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
C4H12KNaO10
MFCD00150989
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SDS Documents