missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Sodium L-(+)-Tartrate Tetrahydrate, TCI America™

Manufacturer:  TCI America P1798100G

 View more versions of this product

Catalog No. P1798100G



Specifications

Specifications

Potassium Sodium L-(+)-Tartrate Tetrahydrate
≥98.0% (T)
MFCD00150989
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
2724148
Crystal-Powder at 20°C
80°C
6381-59-5
C4H12KNaO10
L-(+)-Tartaric Acid Potassium Sodium Salt, Rochelle Salt, Seignette Salt
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
282.218
210.16
White
100g

Chemical Identifiers

6381-59-5
282.218
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
C4H12KNaO10
MFCD00150989
L-(+)-Tartaric Acid Potassium Sodium Salt, Rochelle Salt, Seignette Salt
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents