missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium sodium L-tartrate tetrahydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1016330

 View more versions of this product

Catalog No. AAA1016330



Description

Description

Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Baking powder, medicine (cathartic), component of Fehling′s solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.

Solubility
Soluble in water. Insoluble in alcohol

Notes
Loses water of crystallization at 140°. Unstable above 225°. Incompatible with strong oxidizing agents.
Specifications

Specifications

Potassium sodium L-tartrate tetrahydrate
70°C to 80°C
99%
MFCD00150989
6113568
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
2724148
99%
6381-59-5
1.77
C4H12KNaO10
250 g
14,7670
Soluble in water; Insoluble in alcohol
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
282.218
282.23 (210.29 Anhydrous)

Chemical Identifiers

6381-59-5
282.218
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
C4H12KNaO10
MFCD00150989
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
Safety and Handling

Safety and Handling

EINECSNumber : 206-156-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only