missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium silicate, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 044493A3

 View more versions of this product

Catalog No. AA44493A3



Description

Description

Potassium silicate is used in binders, carbon electrodes, lead pencils, protective coatings, insoluble pigments, phosphors on television tubes and detergent additives. It acts as a catalyst to make glass, ceramics, welding rods, adhesives, cements, gels, paints, pulp and paper production. Further, it is used in horticulture as a soluble source of potassium and silicon. It acts as a corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium silicate
1312-76-1
2 kg
MFCD00145152
Odorless
White
Powder
SiO2:K2O 2.5:1 wt%
Soluble in water.

Chemical Identifiers

1312-76-1
SiO2:K2O 2.5:1 wt%
MFCD00145152
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 215-199-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only