Learn More
Potassium selenocyanate, 99%, Thermo Scientific Chemicals
$143.21 - $530.67
Chemical Identifiers
CAS | 3425-46-5 |
---|---|
Molecular Formula | CKNSe |
Molecular Weight (g/mol) | 144.08 |
MDL Number | MFCD00011366 |
InChI Key | KYEKHFSRAXRJBR-UHFFFAOYSA-M |
Synonym | potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g |
PubChem CID | 76960 |
IUPAC Name | potassium;selenocyanate |
SMILES | C(#N)[Se-].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC201980100
|
Thermo Scientific Chemicals
201980100 |
10 g | Glass bottle |
Each for $143.21
|
|
||||
AC201980500
|
Thermo Scientific Chemicals
201980500 |
50 g | Glass bottle |
Each for $530.67
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3425-46-5 | |
144.08 | |
KYEKHFSRAXRJBR-UHFFFAOYSA-M | |
76960 | |
C(#N)[Se-].[K+] |
CKNSe | |
MFCD00011366 | |
potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g | |
potassium;selenocyanate |
Specifications
152.0°C to 158.0°C | |
Orange to Red | |
10 g | |
CKNSe | |
MFCD00011366 | |
potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g | |
KYEKHFSRAXRJBR-UHFFFAOYSA-M | |
potassium;selenocyanate | |
76960 | |
99% | |
Glass bottle |
3425-46-5 | |
Crystalline Solid, Powder, Crystals and/or Chunks | |
98.5 % min. (Argentometry) | |
KSeCN | |
06,487; 08,417; 13,186; 08,417; 15,274 | |
Solubility in water: soluble | |
C(#N)[Se-].[K+] | |
144.08 | |
144.08 | |
Authentic | |
Potassium selenocyanate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store in a well-
GHS Signal Word: Danger
EINECSNumber : 222-320-1
RUO – Research Use Only