Please login to your online account to display your discounted pricing

Potassium selenocyanate, 99%, ACROS Organics™


Quantity 10g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  201980100

 View more versions of this product

Catalog No. AC201980100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (Argentometry)
Formula Weight 144.08
Physical Form Crystalline Solid, Powder, Crystals and/or Chunks
Quantity 10g
Color Orange to red
Packaging Glass Bottle in Tin Can
Melting Point 152° to 158°C
Fieser 06,487; 08,417; 13,186
Linear Formula KSeCN
Infrared Spectrum Authentic
Assay Percent Range 99%
Chemical Name or Material Potassium selenocyanate
CAS 3425-46-5
Molecular Formula CKNSe
MDL Number MFCD00011366
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

Specifications

Chemical Name or Material Potassium selenocyanate
Assay ≥98.5% (Argentometry)
Assay Percent Range 99%
Molecular Formula CKNSe
Linear Formula KSeCN
Formula Weight 144.08
MDL Number MFCD00011366
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents