missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium selenocyanate, 98.5%

Manufacturer:  Alfa Aesar™ 6810709

 View more versions of this product

Catalog No. AA6810709



Specifications

Specifications

Potassium selenocyanate
CKNSe
UN3283
KYEKHFSRAXRJBR-UHFFFAOYSA-M
potassium;selenocyanate
76960
Crystalline
2.347g/mL
Moisture Sensitive
162°C to 164°C
3425-46-5
MFCD00011366
potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g
C(#N)[Se-].[K+]
144.087
144.08
98.5%
Odorless
White
10g

Chemical Identifiers

3425-46-5
144.087
KYEKHFSRAXRJBR-UHFFFAOYSA-M
76960
C(#N)[Se-].[K+]
CKNSe
MFCD00011366
potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g
potassium;selenocyanate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

RTECSNumber : VT4000000

TSCA : TSCA

SDS Documents