missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium selenate, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 6811118

 View more versions of this product

Catalog No. AA6811118



Specifications

Specifications

Potassium selenate
99.5% (metals basis)
MFCD00054125
YAZJAPBTUDGMKO-UHFFFAOYSA-L
dipotassium;selenate
62680
Powder
White
7790-59-2
K2O4Se
potassium selenate, dipotassium selenate, unii-99798pu7mi, selenic acid, dipotassium salt, 2k.so4e, selenic acid solution, selenic acid, potassium salt 1:2, selenic acid solution, 40 wt. % in water 100g
[O-][Se](=O)(=O)[O-].[K+].[K+]
221.164
221.16
Odorless
50g

Chemical Identifiers

7790-59-2
221.164
YAZJAPBTUDGMKO-UHFFFAOYSA-L
62680
[O-][Se](=O)(=O)[O-].[K+].[K+]
K2O4Se
MFCD00054125
potassium selenate, dipotassium selenate, unii-99798pu7mi, selenic acid, dipotassium salt, 2k.so4e, selenic acid solution, selenic acid, potassium salt 1:2, selenic acid solution, 40 wt. % in water 100g
dipotassium;selenate
Safety and Handling

Safety and Handling

H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

EINECSNumber : 232-214-7

SDS Documents