Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium selenate, 99%

Chemical Identifiers

Chemical Name or Material Potassium selenate
CAS 7790-59-2
Molecular Formula K2SeO4
Formula Weight 221.16
MDL Number MFCD00054125
UN Number UN2630
Merck Index 14,7666
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1613509

  SDS
alfa aesar™
A1613509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.16
UN Number UN2630
Health Hazard 2 GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
MDL Number MFCD00054125
Assay 99%
CAS 7790-59-2
Odor Odorless
Chemical Name or Material Potassium selenate
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Merck Index 14,7666
Health Hazard 1 Danger
Molecular Formula K2SeO4
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENATES
TSCA TSCA
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.