Please login to your online account to display your discounted pricing

Potassium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SP351500

 View more versions of this product

Catalog No. SP351-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 0°C
Purity Grade Certified Grade
Boiling Point 100°C
Quantity 500mL
Physical Form Liquid
Color Colorless
Density 1.0g/cm³
Vapor Pressure 14mmHg at 20°C
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Traceability to NIST LOT NUMBER USED POTASSIUM REF. STD.
CAS 7447-40-7
MDL Number MFCD00133776
ChemAlert Storage Symbol Gray
Packaging Poly Bottle
CAS 7732-18-5
Chemical Name or Material Potassium, Reference Standard Solution
Concentration 990 to 1010ppm
Name Note 1000ppm ±1%

Specifications

Chemical Name or Material Potassium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7447-40-7
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00133776
Accessories
 

SDS & Safety Documents