missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
$123.46 - $126.47

Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.548 |
MDL Number | MFCD00133776 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
IUPAC Name | potassium;chloride |
SMILES | [Cl-].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
SP351-100
|
Fisher Chemical
FLSP351100 |
100mL | Poly Bottle |
Each for $123.46
Case of 6 Each for $740.76 |
|
SP351-500
|
Fisher Chemical
SP351500 |
500mL | Poly Bottle |
Each for $126.47
Case of 6 Each for $758.82 |
|
Chemical Identifiers
7447-40-7 | |
74.548 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
CHEBI:32588 | |
[Cl-].[K+] |
ClK | |
MFCD00133776 | |
4873 | |
potassium;chloride |
Specifications
1000ppm ±1% | |
ClK | |
990 to 1010ppm | |
[Cl-].[K+] | |
74.548 | |
CHEBI:32588 | |
Certified | |
LOT NUMBER USED POTASSIUM REF. STD. | |
1.0g/cm³ | |
100°C | |
100mL |
7447-40-7,7732-18-5 | |
MFCD00133776 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
Liquid | |
Poly Bottle | |
14mmHg at 20°C | |
0°C | |
Colorless | |
Potassium, Reference Standard Solution, 1000ppm ±1% |
Safety and Handling
