Please login to your online account to display your discounted pricing

Potassium Pyrosulfate, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  2964-01

 View all options for this product

Catalog No. 02003828

 

Description & Specifications

Chemical Identifiers

Packaging Wide Mouth Amber Glass Bottle
Quantity 500g
Specific Gravity 1L = 2.28kg
Quantity 500g
Chemical Name or Material Potassium Pyrosulfate
CAS 7790-62-7
Purity Grade BAKER ANALYZED™ Reagent

Specifications

Chemical Name or Material Potassium Pyrosulfate
CAS 7790-62-7
Purity Grade BAKER ANALYZED™ Reagent