Please login to your online account to display your discounted pricing

Potassium Pyrosulfate, ACS Grade, LabChem™

$285.09 - $285.09

Chemical Identifiers

Chemical Name or Material Potassium Pyrosulfate
CAS 7790-62-7
CAS Max % 100
Purity Grade ACS Grade
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC200981

 
lab chem
LC200981
500g Each for $285.09
Description & Specifications

Specifications

Chemical Name or Material Potassium Pyrosulfate
Identification Passes Test
Packaging Amber Glass
Purity Grade ACS Grade
Physical Form Powder
Solubility Soluble in water
Storage Information Room Temperature
CAS 7790-62-7
UN Number UN3260
Decomposition Information Sulfur compounds
Color White
Melting Point 325°C
Quantity 500g
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Keep only in original conatainer.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Danger

GHS P Statement
Keep only in original conatainer.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause respiratory irritation.