Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium pyrophosphate, 95%

Chemical Identifiers

Chemical Name or Material Potassium pyrophosphate
CAS 7320-34-5
Formula Weight 330.34
MDL Number MFCD00011393
Merck Index 14,7663
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13436A1

 
alfa aesar™
13436A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13436A7

 
alfa aesar™
13436A7
5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 330.34
Chemical Name or Material Potassium pyrophosphate
TSCA TSCA
Physical Form Powder
Assay 95%
Sensitivity Hygroscopic
MDL Number MFCD00011393
CAS 7320-34-5
Merck Index 14,7663
Color White
Melting Point 1090°C
Quantity 1kg
Solubility Information Soluble in water
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.33g/mL
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.