missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium, plasma standard solutionμg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 14379AP

 View more versions of this product

Catalog No. AA14379AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium
Clear liquid
MFCD00133776
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
Liquid
Colorless
7757-79-1,7697-37-2,7732-18-5
KNO3
Nitric acid: 5.0%; Potassium nitrate: 2.6%; Water: 92.4%
[N+](=O)([O-])[O-].[K+]
101.102
CHEBI:63043
23 hPa (17mm Hg) at 20°C
500mL

Chemical Identifiers

7757-79-1
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
[N+](=O)([O-])[O-].[K+]
KNO3
MFCD00133776
24434
potassium;nitrate
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents