missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13866AD

 View more versions of this product

Catalog No. AA13866AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium
Clear liquid
MFCD00133776
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
Liquid
Colorless
7757-79-1,7757-79-1,7732-18-5
KNO3
Nitric acid: 5.0%; Potassium nitrate: 0.26%; Water: 94.74%
[N+](=O)([O-])[O-].[K+]
101.102
CHEBI:63043
23 hPa (17mm Hg) at 20°C
50mL

Chemical Identifiers

7757-79-1
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
[N+](=O)([O-])[O-].[K+]
KNO3
MFCD00133776
24434
potassium;nitrate
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

SDS Documents