Please login to your online account to display your discounted pricing

Potassium Phthalate ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 12kg
Packaging STAKMOR

Manufacturer: avantor performance  670420

 View more versions of this product

Catalog No. MK6704A12

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging STAKMOR
Quantity 12kg
Chemical Name or Material Potassium Phthalate
Name Note Primary Standard
CAS 877-24-7
Purity Grade ACS
Physical Form Crystalline
Molecular Formula 1-KOCOC6H4-2-COOH
Formula Weight 204.22g/mol

Specifications

Chemical Name or Material Potassium Phthalate
Name Note Primary Standard
CAS 877-24-7
Purity Grade ACS
Physical Form Crystalline
Molecular Formula 1-KOCOC6H4-2-COOH
Formula Weight 204.22g/mol