missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium phosphate, tribasic monohydrate, 96%, extra pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC263790251

 View more versions of this product

Catalog No. AC263790251

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium phosphate, tribasic monohydrate
27176-10-9
K3O4P·H2O
MFCD00150410
Solubility in water: soluble. Other solubilities: insoluble in alcohol
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
230.28
230.28
96%
50ppm max
2.564
20ppm max.
Fibre drum
200ppm max.
1340.0°C
c.p.
96%
K3PO4·H2O
acmc-1chx7
RMNIZOOYFMNEJJ-UHFFFAOYSA-K
tripotassium;phosphate;hydrate
22031701
Powder
Extra Pure
50ppm max.
20ppm max.
13% max. (800°C)
Solubility in water: soluble. Other solubilities: insoluble in alcohol
White
25kg

Chemical Identifiers

27176-10-9
230.28
RMNIZOOYFMNEJJ-UHFFFAOYSA-K
22031701
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
K3O4P·H2O
MFCD00150410
acmc-1chx7
tripotassium;phosphate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis

GHS Signal Word: Danger

SDS Documents