Please login to your online account to display your discounted pricing

Potassium Phosphate pH 7.5 DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™

$751.80 - $880.78

Chemical Identifiers

Chemical Name or Material Potassium Phosphate pH 7.5 DILUT-IT™ Dissolution Media Concentrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003698

 
avantor performance
D01600
Poly Bottle 230.8mL Each for $880.78

02003699

 
avantor performance
D01602
Narrow Mouth Polyethylene Bottle 961.5mL Each for $751.80
Description & Specifications

Specifications

Chemical Name or Material Potassium Phosphate pH 7.5 DILUT-IT™ Dissolution Media Concentrate
Specific Gravity 1L = 1.00kg
Packaging Poly Bottle
Quantity 230.8mL