Please login to your online account to display your discounted pricing

Potassium Phosphate pH 7.2 DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™

$867.10 - $1,182.71

Chemical Identifiers

Chemical Name or Material Potassium Phosphate pH 7.2 DILUT-IT™ Dissolution Media Concentrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003697

 
avantor performance
D01504
Narrow Mouth Polyethylene Bottle 1.9L Each for $1,182.71

02003696

 
avantor performance
D01502
Narrow Mouth Polyethylene Bottle 961.5mL Each for $867.10
Case of 6 Each for $5,202.60
Description & Specifications

Specifications

Chemical Name or Material Potassium Phosphate pH 7.2 DILUT-IT™ Dissolution Media Concentrate
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.15kg
Quantity 1.9L