Please login to your online account to display your discounted pricing

Potassium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™


Quantity 961.5mL
Packaging Narrow Mouth Polyethylene Bottle

Manufacturer: avantor performance  D01402

 View more versions of this product

Catalog No. 02003695

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Potassium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.10kg
Quantity 961.5mL

Specifications

Chemical Name or Material Potassium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate