Please login to your online account to display your discounted pricing

Potassium Phosphate pH 5.8 DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™

$735.93 - $1,004.44

Chemical Identifiers

Chemical Name or Material Potassium Phosphate pH 5.8 DILUT-IT™ Dissolution Media Concentrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003690

 
avantor performance
D01202
Narrow Mouth Polyethylene Bottle 961.5mL Each for $735.93

02003691

 
avantor performance
D01204
Narrow Mouth Polyethylene Bottle 1.9L Each for $1,004.44

02003692

 
avantor performance
D01200
Poly Bottle 230.8mL Each for $823.68
Description & Specifications

Specifications

Chemical Name or Material Potassium Phosphate pH 5.8 DILUT-IT™ Dissolution Media Concentrate
Quantity 961.5mL
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.11kg