Please login to your online account to display your discounted pricing

Potassium Phosphate pH 5.8 DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™


Quantity 1.9L
Packaging Narrow Mouth Polyethylene Bottle

Manufacturer: avantor performance  D01204

 View more versions of this product

Catalog No. 02003691

  • / Each
 

Description & Specifications

Chemical Identifiers

Packaging Narrow Mouth Polyethylene Bottle
Quantity 1.9L
Chemical Name or Material Potassium Phosphate pH 5.8 DILUT-IT™ Dissolution Media Concentrate
Specific Gravity 1L = 1.11kg

Specifications

Chemical Name or Material Potassium Phosphate pH 5.8 DILUT-IT™ Dissolution Media Concentrate