Please login to your online account to display your discounted pricing

Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, EMD Millipore

$48.75 - $113.00

Chemical Identifiers

Assay Percent Range ≥99%
Formula Weight 136.1
CAS 7778-77-0
Chemical Name or Material Potassium Phosphate, Monobasic
Molecular Formula KH2PO4
Purity Grade Notes by titration
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5295681KG

 
emd millipore corporation
5295681KG
Each for $113.00

529568250GM

 
emd millipore corporation
529568250GM
Each for $48.75
Description & Specifications

Specifications

Formula Weight 136.1
Quantity 1kg
Purity Grade Notes by titration
Molecular Formula KH2PO4
CAS 7778-77-0
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
DNase None detected
RTECS Number TC6615500
Chemical Name or Material Potassium Phosphate, Monobasic
EINECS Number 231-913-4
Purity Molecular biology grade
RNase None detected
Assay Percent Range ≥99%
Safety and Handling

Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).