missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Phosphate Monobasic (Crystalline/NF/FCC), Fisher Chemical

Manufacturer:  Thermo Scientific FLP380250LB

 View more versions of this product

Catalog No. P380250LB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium Phosphate Monobasic
FCC/NF
H2KO4P
3mg/kg max. (FCC), 3ppm max. (NF)
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium;dihydrogen phosphate
516951
136.09
2.34g/mL
0.0015% max. (FCC), 0.0015% max. (FCC), 0.002% max. (NF), 0.002% max. (NF)
0.2% max. (FCC), 0.2% max. (FCC/NF), 0.2% max. (NF)
1% max. (FCC), 1% max. (FCC), 1.0% max. (NF), 1.0% max. (NF)
Pass Test (NF)
White
4.5
7778-77-0
≥98.0% (FCC)
MFCD00011401
Potassium Acid Phosphate, Potassium Biphosphate, Potassium Dihydrogen Phosphate
OP(=O)(O)[O-].[K+]
136.084
CHEBI:63036
Solid
0.001% max. (NF), 10mg/kg max. (FCC), 10mg/kg max. (FCC), 0.001% max. (NF)
Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
2mg/kg max. (FCC), 2mg/kg max. (FCC), 5ppm max. (NF), 5ppm max. (NF)
Fiber Drum
>250°C
253°C
250 lb.

Chemical Identifiers

7778-77-0
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
potassium;dihydrogen phosphate
H2KO4P
MFCD00011401
Potassium Acid Phosphate, Potassium Biphosphate, Potassium Dihydrogen Phosphate
CHEBI:63036
OP(=O)(O)[O-].[K+]
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1

SDS Documents