Please login to your online account to display your discounted pricing

Potassium Phosphate, Monobasic, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$212.75 - $688.35

Chemical Identifiers

Chemical Name or Material Potassium Phosphate
CAS 7778-77-0
Name Note Monobasic
Purity Grade Baker Analyzed™ ACS Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula KH2PO4
Formula Weight 136.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003816

 
avantor performance
324601
Poly Bottle 500g Each for $212.75

02003817

 
avantor performance
324605
Poly bottle 2.5kg Each for $688.35
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 136.09
Purity Grade Notes Meets ACS reagent chemical requirements
Chemical Name or Material Potassium Phosphate
Molecular Formula KH2PO4
CAS 7778-77-0
Name Note Monobasic
Packaging Poly Bottle
Specific Gravity 1L = 2.34kg
Quantity 500g