Please login to your online account to display your discounted pricing

Potassium Phosphate, Monobasic, ACS Grade, ≥ 99.0%, LabChem™

$96.94 - $96.94

Chemical Identifiers

Chemical Name or Material Potassium Phosphate
CAS 7778-77-0
CAS Max % 100
Name Note Monobasic
Purity Grade ACS Grade
Linear Formula KH2PO4
Formula Weight 136.09
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC200951

 
lab chem
LC200951
500g Each for $96.94
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 136.09
Chemical Name or Material Potassium Phosphate
EINECS Number 231-913-4
Identification Passes Test
Name Note Monobasic
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula KH2PO4
CAS 7778-77-0
Decomposition Information Phosphorus oxides; Phosphine
Melting Point 253°C
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Color White
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.