Please login to your online account to display your discounted pricing

Potassium phosphate, monobasic, 99+%, extra pure, ACROS Organics™

$108.17 - $108.17

Chemical Identifiers

Chemical Name or Material Potassium phosphate, monobasic
CAS 7778-77-0
Purity Grade Extra Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula H2KO4P
Linear Formula KH2PO4
Formula Weight 136.09
MDL Number MFCD00011401
Synonym Potassium dihydrogen phosphate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205920100

  SDS
thermo fisher scientific
AC205920100
Plastic Bucket 10kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC205920025

  SDS
fisher scientific acros
205920025
Plastic bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC205925000

  SDS
fisher scientific acros
205925000
Plastic bottle 500g Each for $108.17
Description & Specifications

Specifications

Purity Grade Extra Pure
Physical Form Crystals or Crystalline Powder
Assay ≥99.0%
Formula Weight 136.09
Quantity 10kg
Color Colorless to white
Packaging Plastic Bucket
pH 4.2 to 4.7 (1% soln. at 20°C)
Merck Index 15, 7780
Chemical Name or Material Potassium phosphate, monobasic
Molecular Formula H2KO4P
Linear Formula KH2PO4
Sulfate 100ppm max.
CAS 7778-77-0
Synonym Potassium dihydrogen phosphate
Chlorine (Cl) 30ppm max.
Heavy Metals (as Pb) 10ppm max.
Loss on Drying 0.4% max. (105°C)
Assay Percent Range 99+%
MDL Number MFCD00011401