missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium phosphate, monobasic, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205920025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium phosphate, monobasic | |
7778-77-0 | |
4.2 to 4.7 (1% soln. at 20°C) | |
2.5 kg | |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 | |
136.09 | |
Extra Pure | |
Plastic bottle |
253.0°C | |
Colorless to White | |
Crystalline Powder and/or Crystals | |
99+% | |
KH2PO4 | |
15, 7780 | |
Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 | |
≥99% | |
0.4% max. (105°C) |
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
potassium dihydrogen phosphate |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
CHEBI:63036 | |
[K+].OP(O)([O-])=O |
Safety and Handling
EINECSNumber : 231-913-4
RUO – Research Use Only