missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Phosphate Monobasic, 99+%, (Crystalline), MP Biomedicals
Supplier: MP Biomedicals Inc 0219545301
Specifications
Potassium Phosphate Monobasic | |
7778-77-0 | |
1 kg | |
H2KO4P | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 |
Anhydrous | |
Crystalline | |
99+% | |
MFCD00011401 MFCD00147253 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 | |
136.09 |
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
potassium dihydrogen phosphate |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
CHEBI:63036 | |
[K+].OP(O)([O-])=O |
Safety and Handling
Recommended Storage : Room Temperature